Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human Chromogranin A, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569898
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human samples. It is validated for Intracellular staining (flow cytometry)
Specifications
Chromogranin A | |
Monoclonal | |
Brilliant Violet 421 | |
CGA; CgA; CHGA; Chromogranin-A; CMGA; SP-I | |
Human Chromogranin A Recombinant Protein | |
25 Tests | |
1113 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
S21-537 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction