Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human IL-33 Receptor (ST2), BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | IL-33 Receptor (ST2) |
---|---|
Clone | U33-374 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 421 |
Description
IL-33 Receptor (ST2) Monoclonal antibody specifically detects IL-33 Receptor (ST2) in Human samples. It is validated for Flow cytometrySpecifications
IL-33 Receptor (ST2) | |
Flow Cytometry | |
Brilliant Violet 421 | |
Mouse | |
Human | |
IL1RL1; IL33R; ST2; interleukin-1 receptor-like 1 | |
Human IL-33R Recombinant Protein | |
Affinity Chromatography |
U33-374 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
9173 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title