Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Human Pre-BCR, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570135
Description
Pre-BCR Monoclonal antibody specifically detects Pre-BCR in Human samples. It is validated for Flow cytometry|Intracellular staining (flow cytometry)
Specifications
| Pre-BCR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Liquid |
| Flow Cytometry | |
| HSL2 | |
| Brilliant Violet 421 | |
| Pre-B Cell Receptor | |
| Human Pre-BCR Complexes Purified from Ltk-Pre-BCR Transfectants | |
| 25 μg | |
| 613 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction