Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Mouse Anti-Human TIGIT, BD Horizon™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 570445

 View more versions of this product

Catalog No. BDB570445


Only null left
Add to Cart

Description

Description

TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIGIT
Monoclonal
Brilliant Violet 421
T-cell immunoreceptor with Ig and ITIM domains; VSIG9; VSTM3; WUCAM
Human TIGIT Transfected Cell Line
25 Tests
201633
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
IgG1 κ
Flow Cytometry
TgMab-2
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
Affinity Chromatography
RUO
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.