Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Mouse Anti-Mouse CD369 (CLEC7A), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569709
Description
CD369 (CLEC7A) Monoclonal antibody specifically detects CD369 (CLEC7A) in Mouse samples. It is validated for Flow cytometry
Specifications
CD369 (CLEC7A) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
2F3 | |
Brilliant Violet 421 | |
Clec7a; CD369; BGR; Beta-glucan receptor; Bgr; Dectin-1; Dectin1; dectin-1 | |
Mouse Dectin-1 Recombinant Protein | |
0.1 mg | |
56644 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction