Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Mouse Anti-Rat Integrin αE2 (CD103), BD OptiBuild™

Catalog No. BDB744817 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744817 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744817 Supplier BD Biosciences Supplier No. 744817
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin αE2 (CD103) Monoclonal antibody specifically detects Integrin αE2 (CD103) in Rat samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αE2 (CD103)
Applications Flow Cytometry
Classification Monoclonal
Clone OX-62
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin alpha E; Itgae; Integrin αE; CD103
Host Species Mouse
Immunogen Density gradient-enriched PVG rat thoracic-duct dendritic cells
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV421 under optimal conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.