Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Rat Anti-Human CAT1 (SLC7A1), BD Horizon™

Catalog No. BDB570047 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570047 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570047 Supplier BD Biosciences Supplier No. 570047
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CAT1 (SLC7A1) Monoclonal antibody specifically detects CAT1 (SLC7A1) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAT1 (SLC7A1)
Applications Flow Cytometry
Classification Monoclonal
Clone CA2
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ATRC1; CAT-1; ERR; REC1L; SLC7A1; ecotropic retroviral leukemia receptor homolog
Host Species Rat
Immunogen Human CAT1 Transfected Cells
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 6541
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.