Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV421 Rat Anti-Human Folate Receptors α/β (FRαβ), BD Horizon™
Rat Monoclonal Antibody
$214.00 - $535.00
Specifications
| Antigen | Folate Receptors α/β (FRαβ) |
|---|---|
| Clone | 5 (also known as No.5; No. 5; Antibody No.5; Antibody No. 5; Clone 5; 5/FRAB) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
Description
Folate Receptors α/β (FRαβ) Monoclonal antibody specifically detects Folate Receptors α/β (FRαβ) in Human samples. It is validated for Flow cytometrySpecifications
| Folate Receptors α/β (FRαβ) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Rat | |
| Human | |
| FOLR1, Folate receptor 1, FR-alpha, FRalpha; FOLR2, Folate receptor 2, FR-beta, FRbeta; FRαβ, FR-αβ | |
| Human Folate Receptor beta Transfected Cell Line | |
| Affinity Chromatography |
| 5 (also known as No.5; No. 5; Antibody No.5; Antibody No. 5; Clone 5; 5/FRAB) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 2348, 2350 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title