Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Rat Anti-Mouse CD140b (PDGFRβ), BD Horizon™

Catalog No. BDB570045 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570045 0.1 mg
BDB570126 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570045 Supplier BD Biosciences Supplier No. 570045
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD140b (PDGFRβ) Monoclonal antibody specifically detects CD140b (PDGFRβ) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD140b (PDGFRβ)
Applications Flow Cytometry
Classification Monoclonal
Clone APB5
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD140b; PDGF-R-beta; PDGFR-beta; Pdgfrb; platelet-derived growth factor receptor beta
Host Species Rat
Immunogen Recombinant Mouse PDGFRβ Extracellular Domain Protein
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 18596
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.