Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Rat Anti-Mouse CD8b, BD Horizon™

Rat Monoclonal Antibody

Supplier:  BD Biosciences 569947

 View more versions of this product

Catalog No. BDB569947


Only null left
Add to Cart

Description

Description

CD8b Monoclonal antibody specifically detects CD8b in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD8b
Monoclonal
0.2 mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
Affinity Chromatography
RUO
Mouse
Liquid
Flow Cytometry
YTS156.7.7.rMAb (also known as YTS156.7.7; YTS156)
Brilliant Violet 421
Cd8b1; Cd8b; Ly-3; Lyt-3; lymphocyte antigen 3
Mouse Thymocytes
25 μg
12526
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.