Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV421 Rat Anti-Mouse TCR Vβ14, BD Horizon™

Catalog No. BDB568788 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568788 0.1 mg
BDB568789 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB568788 Supplier BD Biosciences Supplier No. 568788
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TCR Vβ14 Monoclonal antibody specifically detects TCR Vβ14 in Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR Vβ14
Applications Flow Cytometry
Classification Monoclonal
Clone 14-2
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias TCR Vbeta14; TCR Vβ 14; Tcrb-V14; Vβ 14 TCR
Host Species Rat
Immunogen Mouse B10.A Helper T-Cell Clone J9.19
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 110361
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.