Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV480 Mouse Anti-Human Neuropilin-1 (CD304), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756733
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry
Specifications
Neuropilin-1 (CD304) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
mAb3 (also known as MAB3; mAb#3) | |
Brilliant Violet 480 | |
BDCA-4; Blood dendritic cell antigen 4; NRP; NRP1; Neuropilin; VEGF165R | |
Human NRP1 b1b2 Ectodomain | |
50 μg | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction