Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV480 Mouse Anti-Human PD-L2 (CD273), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569906
Description
PD-L2 (CD273) Monoclonal antibody specifically detects PD-L2 (CD273) in Human samples. It is validated for Flow cytometry
Specifications
PD-L2 (CD273) | |
Monoclonal | |
Brilliant Violet 480 | |
CD273; PD-L2; PDL2; PDCD1LG2; B7-DC; B7DC | |
Human PD-L2 cDNA | |
25 Tests | |
80380 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Flow Cytometry | |
24F.10C12 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction