Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV480 Mouse Anti-Human TCR Vα24, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 758079
Description
TCR Vα24 Monoclonal antibody specifically detects TCR Vα24 in Human samples. It is validated for Flow cytometry
Specifications
TCR Vα24 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
C15 | |
Brilliant Violet 480 | |
TCR Va24; TCR Valpha24; TCR Vα24; TCRAV24S1; TCRV alpha 24; TRAV24 | |
Human T-cell clone | |
50 μg | |
28659 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction