Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV480 Mouse Anti-Human TIGIT, BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | TIGIT |
---|---|
Clone | TgMab-2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 480 |
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow cytometrySpecifications
TIGIT | |
Flow Cytometry | |
Brilliant Violet 480 | |
Mouse | |
Human | |
T-cell immunoreceptor with Ig and ITIM domains; VSIG9; VSTM3; WUCAM | |
Human TIGIT Transfected Cell Line | |
Affinity Chromatography |
TgMab-2 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
201633 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title