Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV510 Mouse Anti-Human PD-L2 (CD273), BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $486.00
Specifications
Antigen | PD-L2 (CD273) |
---|---|
Clone | 24F.10C12 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 510 |
Description
PD-L2 (CD273) Monoclonal antibody specifically detects PD-L2 (CD273) in Human samples. It is validated for Flow cytometrySpecifications
PD-L2 (CD273) | |
Flow Cytometry | |
Brilliant Violet 510 | |
Mouse | |
Human | |
CD273; PD-L2; PDL2; PDCD1LG2; B7-DC; B7DC | |
Human PD-L2 cDNA | |
Affinity Chromatography |
24F.10C12 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
80380 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title