Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV510 Rat Anti-Mouse CD39, BD Horizon™

Catalog No. BDB570194 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570194 0.1 mg
BDB570195 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570194 Supplier BD Biosciences Supplier No. 570194
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD39 Monoclonal antibody specifically detects CD39 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39
Applications Flow Cytometry
Classification Monoclonal
Clone Y23-1185
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ectonucleoside triphosphate diphosphohydrolase 1; Entpd1; ecto-ATP diphosphohydrolase 1; ecto-ATPDase 1; ecto-ATPase 1; Cd39; NTPDase 1
Host Species Rat
Immunogen Mouse CD39 Transfected Cell Line
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 12495
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.