Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BV605 Rat Anti-Mouse TNF, BD Horizon™

Catalog No. BDB569297 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569297 25 μg
BDB569296 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569297 Supplier BD Biosciences Supplier No. 569297
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TNF Monoclonal antibody specifically detects TNF in Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Applications Flow Cytometry
Classification Monoclonal
Clone MP6-XT22
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Tnf; Tnfa; TNF alpha; TNF-a; Tnfsf1a; Tnfsf2; TNFSF2; Cachectin; DIF
Host Species Rat
Immunogen Recombinant Mouse TNF
Purification Method Affinity Chromatography
Quantity 25 μg
Regulatory Status RUO
Gene ID (Entrez) 21926
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.