Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV711 Biosimilar Anti-Human CD52 , BD OptiBuild™
Biosimilar Monoclonal Antibody
Supplier: BD Biosciences 758150
Description
CD52 Monoclonal antibody specifically detects CD52 in Human samples. It is validated for Flow cytometry
Specifications
| CD52 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Liquid |
| Flow Cytometry | |
| Alemtuzumab297.rMAb (also known as Alemtuzumab N297A Biosimilar.rMAb) | |
| Brilliant Violet 711 | |
| CAMPATH-1 antigen; HE5 | |
| Human CD52 | |
| 50 μg | |
| 1043 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction