Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV711 Rat Anti-Mouse CD140b (PDGFRβ), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756657
Description
CD140b (PDGFRβ) Monoclonal antibody specifically detects CD140b (PDGFRβ) in Mouse samples. It is validated for Flow cytometry
Specifications
CD140b (PDGFRβ) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Flow Cytometry | |
APB5 | |
Brilliant Violet 711 | |
CD140b; PDGF-R-beta; PDGFR-beta; Pdgfrb; platelet-derived growth factor receptor beta | |
Recombinant Mouse PDGFRβ Extracellular Domain Protein | |
50 μg | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction