Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV750 Rat Anti-Mouse C1q, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756666
Description
C1q Monoclonal antibody specifically detects C1q in Mouse samples. It is validated for Flow cytometry
Specifications
C1q | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
RmC7H8 | |
Brilliant Violet 750 | |
Complement C1q; Complement Component 1q; Complement protein C1q | |
Mouse C1q | |
50 μg | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction