Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BV786 Rabbit Anti-Active Caspase-3, BD Horizon™
Rabbit Monoclonal Antibody
$214.00 - $535.00
Specifications
| Antigen | Active Caspase-3 |
|---|---|
| Clone | C92-605.rMAb |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
Active Caspase-3 Monoclonal antibody specifically detects Active Caspase-3 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
| Active Caspase-3 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 836, 12367 | |
| IgG κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| C92-605.rMAb | |
| Flow Cytometry | |
| Brilliant Violet 786 | |
| Rabbit | |
| Human, Mouse | |
| Apopain; CASP-3; CASP3; CC3; CPP-32; CPP32; Caspase 3; Yama | |
| Human Active Caspase-3 Fragment | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title