Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BVDV Envelope glycoprotein E2 Humanized Recombinant Human Monoclonal Antibody (SAA2087)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA554254

Description
Compatible with Direct ELISA.
The BVDV Envelope glycoprotein E2 mediates viral attachment and entry into host cells by binding to cellular receptors.
Specifications
BVDV Envelope glycoprotein E2 Humanized | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Human | |
Protein A/G | |
RUO | |
Virus | |
Antibody | |
VHH-hFc |
ELISA | |
SAA2087 | |
Unconjugated | |
Envelope glycoprotein E2 | |
Recombinant BVDV Envelope glycoprotein E2. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction