Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BVES Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300006F
Description
BVES Polyclonal specifically detects BVES in Human samples. It is validated for ELISA.Specifications
BVES | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
11149 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
blood vessel epicardial substance, H, HBVES, POP1MGC42413, POPDC1popeye domain containing 1, Popeye domain-containing protein 1, Popeye protein 1 | |
Produced in rabbits immunized with purified, recombinant BVES (NP_671488.1; Met1-Asn36). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction