Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BVT 948

Catalog No. 217610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-761-0 10 mg
21-765-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-761-0 Supplier Tocris Bioscience™ Supplier No. 2176/10
This item is not returnable. View return policy

Non-competitive protein tyrosine phosphatase inhibitor

BVT 948 is a non-competitive, cell-permeable inhibitor of protein tyrosine phosphatases (PTPs) (IC50 = 0.09 - 1.7 μM); displays irreversible inhibition through catalysis of the hydrogen peroxide-dependent oxidation of PTP. Enhances insulin signaling in vitro and insulin tolerance in ob/ob mice in vivo. Also inhibits several cytochrome P450 isoforms (IC50 <10 μM).

Specifications

Quantity 10 mg
Product Type BVT 948
Purity >98%
Molecular Weight (g/mol) 241.25
Inhibitors BVT 948

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.