Learn More
Honeywell BW™ Clip Multi-Gas Portable Gas Detector
Catalog No. 19807570
Click to view available options
Battery Life:
2 Years, continuous operation
Catalog No. Battery Life
19-807-570 2 Years, continuous operation
1 options
Catalog No. 19-807-570 Supplier Honeywell Supplier No. BWC4YN
BW™ Clip Multi-Gas Portable Gas Detector is a low maintenance multi-gas detector that requires no charging and no battery or sensor servicing.
  • Simple one-button operation
  • User-friendly, tamper-proof operation with just one button
  • Intuitive LCD icons
  • Real-time gas concentrations shown on LCD
  • 2 year continuous runtime
  • Includes:
    • Specified sensor
    • Stainless steel alligator clip
    • Concussion-proof housing
    • Test cap
    • Instructions
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 2 Years, continuous operation
Battery Type Lithium
Certifications/Compliance CSA (CSA18CA70205765); Classified by CSA to both US and Canadian Standards as intrinsically safe for Class I, Division 1, Group A, B, C, D and Class I, Zone 0, Group IIC, −40°C ≤ Tamb ≤ 55°C; ATEX (Sira 18ATEX2287X ); II 1G, Ex ia IIC T4 Ga, −40°C ≤ Tamb ≤ 55°C; CE European Conformity
Detection Limits H2S: 0 to 100.0 ppm with 1/0.1 increments; CO: 0 to 1000 ppm with 1 increments; O2: 0 to 25.0% Vol with 0.1 increments; Combustible gases: 0 to 100% LEL (or 0 to 5.0% Vol) with 1 increments
For Use With (Equipment) IntelliDoX Docking System
Height (Metric) 12 cm
Length (Metric) 6.8 cm
Weight (Metric) 233 g
Width (Metric) 3.2 cm