Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell BW™ MicroClip XL, %LEL, O2, H2S, CO

Catalog No. 19807571
Encompass
Encompass_Preferred
Change view
Click to view available options
Components Measured:
Oxygen, Hydrogen Sulfide, Carbon Monoxide, LEL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-807-571 Oxygen, Hydrogen Sulfide, Carbon Monoxide, LEL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-807-571 Supplier Honeywell Supplier No. MCXLXWHMYNA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Honeywell BW™ GasAlertMicroClip provides affordable protection from atmospheric gas hazards and extended battery life, especially in cold weather.

  • Features visual compliance at a glance with the flashing, green IntelliFlash™
  • Easy, one-button operation reduces training time and lets workers focus on the job at hand
  • Simple, cost-effective management of records, calibration and bump testing
  • Easy to wear, slim and compact
  • For standard operation with 2-year warranty
  • Runs an entire shift — even at -4°F (-20°C)
  • IP68 rating — unmatched water protection
  • Compact, lightweight and easy to use — with one-button operation and no extensive training
  • Device management with Honeywell Safety Suite.
  • Standard Package Contents:
    • Detector complete with specified sensor(s), stainless steel alligator clip
      and concussion-proof housing
    • Rechargeable battery
    • Wall outlet charging adaptor
    • Calibration/test cap and hose
    • Manual
    • Multi-language CD manual
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CSA: Class I, Div. 1, Gr. A, B, C, D; ATEX: , II 1 G. Ex ia IIC T4 Ga; IECEx: Ex ia IIC T4 Ga; European Conformity
Components Measured Oxygen, Hydrogen Sulfide, Carbon Monoxide, LEL
Type Multigas Detector
Height (Metric) 11.3 cm
Length (Metric) 6 cm
Weight (Metric) 190 g
Width (Metric) 3.1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.