Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BXDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557286 Supplier Invitrogen™ Supplier No. PA557286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSYKLLLKKS GCRTPRIELE EMGPSLDLVL RRTHLASDDL YKLSMKMPKA LKPKKKKNIS HDTFGTTYGR IHMQKQDLSK LQTRKMKGLK K Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BXDC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rpf2
Gene Accession No. Q9H7B2
Gene Alias 2810470K21Rik; AU040229; AU043242; bA397G5.4; brix domain containing 1; brix domain-containing protein 1; Brix domain-containing protein 1-like protein; BXDC1; homolog of Rpf2; hypothetical protein LOC406587; ik:tdsubc_1a3; MNCb-4285; ribosomal processing factor 2 homolog; ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; ribosome production factor 2 homolog (S. cerevisiae); RP11-397G5.1; Rpf2; Unknown (protein for MGC:134144); wu:fe17d08; wu:fj16d06; xx:tdsubc_1a3; zgc:77150
Gene Symbols Rpf2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BXDC1. Recombinant protein control fragment (Product #RP-95043).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.