Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BXDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570486 Supplier Invitrogen™ Supplier No. PA570486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%; Zebrafish: 93%.

RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BXDC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Rpf2
Gene Accession No. 0
Gene Alias 2810470K21Rik; AU040229; AU043242; bA397G5.4; brix domain containing 1; brix domain-containing protein 1; Brix domain-containing protein 1-like protein; BXDC1; homolog of Rpf2; hypothetical protein LOC406587; ik:tdsubc_1a3; MNCb-4285; ribosomal processing factor 2 homolog; ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; ribosome production factor 2 homolog (S. cerevisiae); RP11-397G5.1; Rpf2; Unknown (protein for MGC:134144); wu:fe17d08; wu:fj16d06; xx:tdsubc_1a3; zgc:77150
Gene Symbols Rpf2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of rat BXDC1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294436
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.