Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BXDC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988558
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8558 Supplier Bioss Supplier No. BS8435R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.

Specifications

Antigen BXDC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rpf2
Gene Accession No. Q9H7B2
Gene Alias 2810470K21Rik; AU040229; AU043242; bA397G5.4; BOS_9618; brix domain containing 1; Brix domain-containing protein 1; BXDC1; homolog of Rpf2; hypothetical protein LOC406587; ik:tdsubc_1a3; LOC100357910; MNCb-4285; ribosomal processing factor 2 homolog; Ribosome biogenesis protein RPF2; Ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; ribosome production factor 2 homolog (S. cerevisiae); RP11-397G5.1; RPF2; rRNA-binding ribosome biosynthesis protein RPF2; wu:fe17d08; wu:fj16d06; xx:tdsubc_1a3; YKR081C; YKR401; zgc:77150
Gene Symbols Rpf2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BXDC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84154
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.