Learn More
Invitrogen™ BXDC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589858
Description
Immunogen sequence: MAATKRKRRG GFAVQAKKPK RNEIDAEPPA KRHATAEEVE EEERDRIPGP VCKGKWKNKE RILIFSSRGI NFRTRHLMQD LRMLMPHSKA DTKMDRKDKL FVINEVCEMK NCNKCIYFEA KKKQDLYMWL SNSPHGPSAK FLVQNIHTLA ELKMTGNCLK GSRPLLSFDP AFDELPHYAL LKELLIQIFS TPRYHPKSQP FVDHVFTFTI LDNRIWFRNF QIIEEDAALV EIGPRFVLNL IKIFQGSFGG PTLYENPHYQ SPNMHRRVIR SITAAKYREK QQVKDVQKLR KKEPKTLLPH DPTADVFVTP AEEKPIEIQW VKPEPKVDLK ARKKRIYKRQ RKMKQRMDSG KTK; Positive Samples: U-87MG, HeLa, A-549, Mouse spleen, Mouse testis, Rat spleen, Rat testis; Cellular Location: Nucleus, nucleolus.
Required for biogenesis of the 60S ribosomal subunit.
Specifications
BXDC2 | |
Polyclonal | |
Unconjugated | |
Brix1 | |
1110064N10Rik; BRIX; brix domain containing 2; brix domain-containing protein 2; Brix1; BRX1, biogenesis of ribosomes; BRX1, biogenesis of ribosomes, homolog; BRX1, biogenesis of ribosomes, homolog (S. cerevisiae); BXDC2; C76935; RGD1308508; ribosome biogenesis protein Brix; ribosome biogenesis protein BRX1 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
294799, 55299, 67832 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2.58 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q4QQT6, Q8TDN6, Q9DCA5 | |
Brix1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-353 of human BRIX1 (NP_0607913). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.