Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BXDC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555136
Description
Immunogen sequence: SGKKSLKRKA AAEELQEAAG AGDGATENGV QPPKAAAFPP GFSISEIKNK QRRHLMFTRW KQQQRKEKLA AKKKLKKERE ALGDKAPPKP VPKTID Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 85%.
BXDC5 may be required for ribosome biogenesis.
Specifications
BXDC5 | |
Polyclonal | |
Unconjugated | |
RPF1 | |
2210420E24Rik; 2310066N05Rik; brix domain containing 5; brix domain-containing protein 5; Bxdc5; Ribosome biogenesis protein RPF1; ribosome production factor 1; ribosome production factor 1 homolog; ribosome production factor 1 homolog (S. cerevisiae); ribosome production factor 1-like protein; RNA processing factor 1; RNA processing factor 1 (RPF1); RP11-118B23.1; RPF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80135 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H9Y2 | |
RPF1 | |
Recombinant protein corresponding to Human BXDC5. Recombinant protein control fragment (Product #RP-94107). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction