Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BYSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588285
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588285 Supplier Invitrogen™ Supplier No. PA588285
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HAEVVVDPED ERAIEMFMNK NPPARRTLAD IIMEKLTEKQ TEVETVMSEV SGFPMPQLDP RVLEVYRGVR EVLSKYRSGK LPKAFKIIPA LSNWEQILYV TEPEAWTAAA MYQATRIFAS NLKERMAQRF YNLVLLPRVR DDVAEYKRLN FHLYMALKKA L; Positive Samples: HeLa, Jurkat, A-549, BxPC-3, Mouse testis, Mouse lung, Rat testis, Rat lung.

Bystin is expressed as a 2-kb major transcript and a 3.6-kb minor transcript in SNG-M cells and in human trophoblastic teratocarcinoma HT-H cells. Protein binding assays determined that bystin binds directly to trophinin and tastin, and that binding is enhanced when cytokeratins 8 and 18 are present. Immunocytochemistry of HT-H cells showed that bystin colocalizes with trophinin, tastin, and the cytokeratins, suggesting that these molecules form a complex in trophectoderm cells at the time of implantation. Using immunohistochemistry it was determined that trophinin and bystin are found in the placenta from the sixth week of pregnancy. Both proteins were localized in the cytoplasm of the syncytiotrophoblast in the chorionic villi and in endometrial decidual cells at the uteroplacental interface. After week 10, the levels of trophinin, tastin, and bystin decreased and then disappeared from placental villi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BYSL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BYSL
Gene Accession No. O54825, Q13895, Q80WL2
Gene Alias by the ribosomal protein s6 gene, drosophila, homolog-like; Bys; BYSL; bysl {ECO:0000312; Bystin; bystin like; bystin-like; EMBL:AAH79030.1, ECO:0000312; Enp1; RGD:727959}
Gene Symbols BYSL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 120-280 of human BYSL (NP_0040443).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359727, 53414, 705
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.