Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bz 423
GREENER_CHOICESDP

Catalog No. 579150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-915-0 50 mg
57-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-915-0 Supplier Tocris Bioscience™ Supplier No. 5791/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

ATP synthase inhibitor; proapoptotic

Bz 423 is an ATP synthase inhibitor (IC50 ~5 μM). Binds the oligomycin sensitivity conferring protein (OSCP) component of mitochondrial F1F0-ATPase in vitro resulting in generation of superoxide and apoptosis. Kills primary B and T cell in mice in vivo. Suppresses autoimmunity and prolongs survival in mouse lupus model.

Specifications

Chemical Name or Material 7-Chloro-1,3-dihydro-5-(4-hydroxyphenyl)-1-methyl-3-(2-naphthalenylmethyl)-2H-1,4-benzodiazepin-2-one
CAS 216691-95-1
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C27H21ClN2O2
Formula Weight 440.92
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.