Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BzATP triethylammonium salt
GREENER_CHOICESDP

Catalog No. 33121 Shop All Novus Products
Change view
Click to view available options
Quantity:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-121 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 33-121 Supplier R&D Systems™ Supplier No. 3312/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Photoaffinity label for ATPase; also P2X7 agonist and P2X1/P2Y1 partial agonist

Prototypic P2X7 receptor agonist (EC50 values are 3.6, 7 and 285 μM for rat, human and mouse receptors respectively). Exhibits 5 - 10 fold greater potency than ATP. Exhibits partial agonist activity at P2X1 (pEC50 = 8.7) and P2Y1 receptors and can be used as a photoaffinity label for ATPase.

This compound is a mixture of isomers.

Chemical Identifiers

CAS 112898-15-4
Molecular Formula C30H39N6O15P3
Synonym 2' 3'-o-4-benzoylbenzoyl adenosine 5'-triphosphate triethylammonium salt

Specifications

Quantity 1mg
Product Line Tocris
Formula Weight 1018.97
Chemical Name or Material BzATP triethylammonium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.