Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BZRAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549434

Catalog No. PA549434


Only null left
Add to Cart

Description

Description

BZRAP1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY
Specifications

Specifications

BZRAP1
Polyclonal
Unconjugated
TSPOAP1
benzodiazapine receptor (peripheral) associated protein 1; benzodiazapine receptor peripheral-associated protein 1; benzodiazepine receptor (peripheral) associated protein 1; BZRAP1; KIAA0612; PBR-IP; Peripheral benzodiazepine receptor-interacting protein; peripheral-type benzodiazepine receptor-associated protein 1; PRAX1; PRAX-1; RBP1; RIMBP1; RIM-BP1; RIMS-binding protein 1; TSPO associated protein 1; TSPOAP1; TSPO-associated protein 1
Rabbit
Antigen affinity chromatography, Protein A
RUO
9256
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O95153
TSPOAP1
KLH conjugated synthetic peptide between 1551-1585 amino acids from the C-terminal region of human BZRAP1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.