Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BZRAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555235
Description
Immunogen sequence: RTASTSTLGE KDPGPAAPSL AKQEAEWTAG EACPASSSTQ GARAQQAPNT EMCQGGDPGS GLRPRAEKED TAELGVHLVN SLVDHGRNSD LSD Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 57%.
Specifications
| BZRAP1 | |
| Polyclonal | |
| Unconjugated | |
| TSPOAP1 | |
| benzodiazapine receptor (peripheral) associated protein 1; benzodiazapine receptor peripheral-associated protein 1; benzodiazepine receptor (peripheral) associated protein 1; BZRAP1; KIAA0612; PBR-IP; Peripheral benzodiazepine receptor-interacting protein; peripheral-type benzodiazepine receptor-associated protein 1; PRAX1; PRAX-1; RBP1; RIMBP1; RIM-BP1; RIMS-binding protein 1; TSPO associated protein 1; TSPOAP1; TSPO-associated protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95153 | |
| TSPOAP1 | |
| Recombinant protein corresponding to Human BZRAP1. Recombinant protein control fragment (Product #RP-93210). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction