Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BZW1 Polyclonal Antibody

Catalog No. PIPA535860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535860 Supplier Invitrogen™ Supplier No. PA535860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (93%), Pig (100%), Chicken (97%), Bovine (100%).

Enhances histone H4 gene transcription but does not seem to bind DNA directly. Target protein's human homolog is involved in transcriptional control at the G1/S phase transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BZW1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Bzw1
Gene Accession No. Q6P7P5, Q7L1Q6, Q9CQC6
Gene Alias 1200015E15Rik; Basic leucine zipper and W2 domain-containing protein 1; basic leucine zipper and W2 domains 1; basic leucine-zipper protein BZAP45; BZAP45; Bzw1; KIAA0005; LOC506562 protein; mKIAA0005; Nbla10236; Protein Orf; putative protein product of Nbla10236
Gene Symbols Bzw1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BZW1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363232, 66882, 9689
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.