Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BZW1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12903R
Description
Enhances histone H4 gene transcription but does not seem to bind DNA directly. Target protein′s human homolog is involved in transcriptional control at the G1/S phase transition.Specifications
| BZW1 | |
| Polyclonal | |
| Unconjugated | |
| Bzw1 | |
| 1200015E15Rik; Basic leucine zipper and W2 domain-containing protein 1; basic leucine zipper and W2 domains 1; basic leucine-zipper protein BZAP45; BOS_1940; BZAP45; BZW1; KIAA0005; LOC100343210; mKIAA0005; Nbla10236; Protein Orf; putative protein product of Nbla10236 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 9689 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q7L1Q6 | |
| Bzw1 | |
| KLH conjugated synthetic peptide derived from human BZW1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction