Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BZW2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554609 Supplier Invitrogen™ Supplier No. PA554609
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLKELSDFLR VQQSLGTRKE LQKELQERLS QECPIKEVVL YVKEEMKRND LPETAVIGLL WTCIMNAVEW NKKEELV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Basic leucine zipper and W2 domain-containing protein 2 (BZW2) may be involved in neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BZW2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BZW2
Gene Accession No. Q91VK1, Q9WTT7, Q9Y6E2
Gene Alias 1110001I24Rik; Basic leucine zipper and W2 domain-containing protein 2; basic leucine zipper and W2 domains 2; Bdm2; Brain development-related molecule 2; BZW2; Hfb2; HSPC028; MST017; MSTP017
Gene Symbols BZW2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BZW2. Recombinant protein control fragment (Product #RP-92731).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171439, 28969, 66912
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.