Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BZW2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100408 Supplier Invitrogen™ Supplier No. PA5100408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BZW2.

Basic leucine zipper and W2 domain-containing protein 2 (BZW2) may be involved in neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BZW2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BZW2
Gene Accession No. Q9Y6E2
Gene Alias 1110001I24Rik; Basic leucine zipper and W2 domain-containing protein 2; basic leucine zipper and W2 domains 2; Bdm2; Brain development-related molecule 2; BZW2; Hfb2; HSPC028; MST017; MSTP017
Gene Symbols BZW2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BZW2(Accession Q9Y6E2), corresponding to amino acid residues C300-C350.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28969
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.