Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BZW2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521808 Supplier Invitrogen™ Supplier No. PA521808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, HeLaS3, Molt-4, Neuro2A, C8D30, Rat-2. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (82%), Pig (99%), Chicken (91%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Basic leucine zipper and W2 domain-containing protein 2 (BZW2) may be involved in neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BZW2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene BZW2
Gene Accession No. Q91VK1, Q9WTT7, Q9Y6E2
Gene Alias 1110001I24Rik; Basic leucine zipper and W2 domain-containing protein 2; basic leucine zipper and W2 domains 2; Bdm2; Brain development-related molecule 2; BZW2; Hfb2; HSPC028; MST017; MSTP017
Gene Symbols BZW2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BZW2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171439, 28969, 66912
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.