Learn More
BZW2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | BZW2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8177
![]() |
Bethyl Laboratories, Inc
A304608AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
Basic leucine zipper and W2 domain-containing protein 2 (BZW2) may be involved in neuronal differentiation.Specifications
BZW2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
HSPC028, MST017, MSTP017, basic leucine zipper and W2 domain-containing protein 2 | |
Between 1 and 50 | |
Primary | |
4° C | |
BZW2 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q91VK1, Q9Y6E2 | |
28969, 66912 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.