Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Abl, p Tyr245 Antibody, Novus Biologicals™
SDP

Catalog No. NB260518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260518 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260518 Supplier Novus Biologicals Supplier No. NBP323313
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

c-Abl Polyclonal antibody specifically detects c-Abl in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen c-Abl
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias Abelson murine leukemia viral oncogene homolog 1, ABL, c-ABL, c-abl oncogene 1, non-receptor tyrosine kinase, c-abl oncogene 1, receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.2, JTK7bcr/abl, p150bcr/c-abl oncogene protein, Proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, tyrosine-protein kinase ABL1, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1
Host Species Rabbit
Immunogen Phospho-c-Abl (Tyr-245) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 245 of human c-Abl. This peptide sequence has high homology to the conserved site in rat and mouse c-Abl, as well as in viral Abl and BCR-Abl fusion protein. [UniProt# P00519]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, DNA Repair, Phospho Specific, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 25
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.