Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. botulinum BoNT-A Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4489SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4489SP 25 μg
MAB4489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4489SP Supplier R&D Systems Supplier No. MAB4489SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BoNT-A Light Chain Monoclonal specifically detects BoNT-A Light Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen BoNT-A Light Chain
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 503013
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q45894
Gene Symbols atx
Host Species Mouse
Immunogen E. coli-derived recombinant Clostridium BoNT-A Light Chain Met1-Phe425 Accession # Q45894
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5185061
Test Specificity Detects C. botulinum BoNT-A Light Chain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with BoNT-B light chain, BoNT-E light chain, BoNT-A heavy chain, BoNT-C1 heavy chain, or BoNT-D heavy chain is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.