Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C. botulinum BoNT-A Light Chain Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4489SP
This item is not returnable.
View return policy
Description
BoNT-A Light Chain Polyclonal specifically detects BoNT-A Light Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BoNT-A Light Chain | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
atx | |
E. coli-derived recombinant Clostridium BoNT-A Light Chain Met1-Phe425 Accession # Q45894 | |
25 μg | |
Primary | |
Detects C. botulinum BoNTA Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant C. botulinum BoNTA Heavy Chain is observed. | |
C. botulinum | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q45894 | |
Sheep | |
Affinity Purified | |
RUO | |
5185061 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction