Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. botulinum BoNT-A Light Chain Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4489
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4489 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4489 Supplier R&D Systems Supplier No. BAF4489
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BoNT-A Light Chain Polyclonal specifically detects BoNT-A Light Chain in C. botulinum samples. It is validated for Western Blot.

Specifications

Antigen BoNT-A Light Chain
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q45894
Gene Symbols atx
Host Species Sheep
Immunogen E. coli-derived recombinant Clostridium BoNTA Light Chain Met1-Phe425 Accession # Q45894
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5185061
Test Specificity Detects C. botulinum BoNTA Light Chain in Western blots. In Western blots, less than 1% crossreactivity with recombinant C. botulinum BoNTA Heavy Chain is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.