Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C. botulinum BoNT-A Light Chain Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548053
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548053 Supplier Invitrogen™ Supplier No. PA548053
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant C. botulinum BoNTA Heavy Chain is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Botulinum neurotoxin type A is one of the seven serotypes of Botulinum Neurotoxins (BoNTs) produced by various strains of Clostridium botulinum. BoNTs are synthesized as inactive single chain protein precursors and activated by proteolytic cleavage to generate disulfide-linked two-chain proteins. The 50 kDa light chain contains the catalytic domain, whereas the 100 kDa heavy chain contains an internal translocation domain and a receptor binding domain. BoNTs are the most potent protein toxins for humans. As zinc proteases, they cleave SNARE proteins to elicit flaccid paralysis in botulism by blocking acetylcholine release at the neuromuscular junction. E. coli-expressed recombinant light chains are active proteases. However, they are not toxic because they cannot enter into host cells in the absence of the heavy chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C. botulinum BoNT-A Light Chain
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative; pH 7.4
Gene Alias BONTC1
Host Species Sheep
Immunogen E. coli-derived recombinant Clostridium BoNT-A Light Chain Met1-Phe425.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.