Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C. botulinum BoNT-B Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5420SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5420SP 25 μg
MAB5420 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5420SP Supplier R&D Systems Supplier No. MAB5420SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BoNT-B Light Chain Monoclonal specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Western Blot.

Specifications

Antigen BoNT-B Light Chain
Applications Western Blot
Classification Monoclonal
Clone 579631
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10844
Gene Alias BoNTB Light Chain, DcD
Gene Symbols boNT/B
Host Species Mouse
Immunogen E. coli-derived recombinant Clostridium BoNT-B Light Chain Pro2-His428 Accession # P10844
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6030102
Test Specificity Detects C. botulinum BoNT-B Light Chain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant Light Chains of BoNT-A, -C1, -D, -E, -F, or -G is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species C. botulinum
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.