Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C. botulinum BoNT-C1 Heavy Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | BoNT-C1 Heavy Chain |
---|---|
Clone | 580223 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
BoNT-C1 Heavy Chain Monoclonal specifically detects BoNT-C1 Heavy Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BoNT-C1 Heavy Chain | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
C. botulinum | |
P18640 | |
10523893 | |
E. coli-derived recombinant C. botulinum BoNT-C1 Heavy Chain Asn866-Glu1291 Accession # P18640 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects C. botulinum BoNT-C1 Heavy Chain in direct ELISAs. In direct ELISAs, no cross-reactivity with the Light Chains of BoNT-A, -B, -C1, -D, -E, -F, -G, or Heavy Chains of BoNT-D or -G is observed. |
580223 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BoNTC1 Heavy Chain, DcD | |
CbC4_4096 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title